missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine ethyl ester hydrochloride, 98%

CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol

Supplier:  Thermo Scientific Chemicals A1361814

Quantity 25 g

 View more versions of this product

Catalog No. AAA1361814


Only null left

Description

Description

L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

Solubility
Soluble in methanol and water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
Specifications

Chemical Identifiers

3182-93-2
229.70
FPFQPLFYTKMCHN-PPHPATTJSA-N
165085
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
C11H16ClNO2
MFCD00012507
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride

Specifications

153°C to 157°C
3182-93-2
MFCD00012507
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
FPFQPLFYTKMCHN-PPHPATTJSA-N
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
229.70
229.71
98%
L-Phenylalanine ethyl ester hydrochloride
C11H16ClNO2
3657823
Soluble in methanol and water.
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
25 g
165085
+36° (c=5 in Ethanol)
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 221-673-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions