missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine ethyl ester hydrochloride, 98%

Catalog No. AAA1361806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1361806 5 g
AAA1361814 25 g
AAA1361822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1361806 Supplier Thermo Scientific Chemicals Supplier No. A1361806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol

L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

Solubility
Soluble in methanol and water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.

Chemical Identifiers

CAS 3182-93-2
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012507
InChI Key FPFQPLFYTKMCHN-PPHPATTJSA-N
Synonym l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
PubChem CID 165085
IUPAC Name ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1

Specifications

Melting Point 153°C to 157°C
Beilstein 3657823
Solubility Information Soluble in methanol and water.
Quantity 5 g
Formula Weight 229.71
Optical Rotation +36° (c=5 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Phenylalanine ethyl ester hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-673-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only