missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Penicillamine, 98%

Catalog No. AC416870050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416870050 5 g Glass bottle
AC416870010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416870050 Supplier Thermo Scientific Chemicals Supplier No. 416870050

CAS: 1113-41-3 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1113-41-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00064303
InChI Key VVNCNSJFMMFHPL-GSVOUGTGSA-N
Synonym l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
PubChem CID 92863
ChEBI CHEBI:50869
IUPAC Name (2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S

Specifications

Melting Point 206.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 5 g
Beilstein 04, III, 1662
Merck Index 15, 7197
Specific Rotation Condition +63° (20°C c=5,1 M NaOH)
Specific Rotation + 63.00
Solubility Information (5% in water) Clear colorless
Formula Weight 149.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Penicillamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 214-203-9
RTECSNumber YV9445500

RUO – Research Use Only