missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Monohydrochloride 98.0+%, TCI America™

Supplier:  TCI America O0064250G

 View more versions of this product

Catalog No. O0064250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-Ornithine Monohydrochloride
White
MFCD00064562
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
C(CC(C(=O)O)N)CN.Cl
168.621
168.62
Crystalline Powder
3184-13-2
C5H13ClN2O2
250 g
GGTYBZJRPHEQDG-WCCKRBBISA-N
(2S)-2,5-diaminopentanoic acid;hydrochloride
76654
≥98.0% (HPLC,T)

Chemical Identifiers

3184-13-2
168.621
GGTYBZJRPHEQDG-WCCKRBBISA-N
76654
C(CC(C(=O)O)N)CN.Cl
C5H13ClN2O2
MFCD00064562
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
(2S)-2,5-diaminopentanoic acid;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1561

RTECSNumber : RM2985000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions