missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride MP Biomedicals

Catalog No. icn10042180
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Catalog No. Quantity
ICN10042180 100 g
1 options

Catalog No. ICN10042180

Supplier: MP Biomedicals Inc 0210042180

Only null left

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Quantity 100 g
Formula Weight 168.6
Chemical Name or Material L-Ornithine Hydrochloride