missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine hydrochloride, 99%

Catalog No. AAA1211136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1211136 500 g
AAA1211114 25 g
AAA1211122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1211136 Supplier Thermo Scientific Chemicals Supplier No. A1211136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3184-13-2 | C5H13ClN2O2 | 168.621 g/mol

L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.

Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point ∼238°C (decomposition)
Beilstein 3625847
Merck Index 14,6874
Solubility Information Soluble in water (543mg/ml at 20°C),and alcohol.
Quantity 500 g
Optical Rotation +23° (c=5 in 5N HCl)
Formula Weight 168.62
Percent Purity 99%
Chemical Name or Material L-Ornithine hydrochloride
EINECSNumber 221-678-6
RTECSNumber RM2985000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only