missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride, 99%, Spectrum™ Chemical

Catalog No. p-7165992
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
25 g
25 kg
5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18611374 25 g Poly Bottle
18611373 100 g Poly Bottle
18611377 500 g Poly Bottle
18611372 1 kg Poly Bottle
18611376 5 kg Poly Pail
18611375 25 kg Fiber Drum
6 options

Catalog No. 18611374

Supplier: Spectrum Chemical Mfg Cor OR13025GM

Only null left

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +23° to +25.0°
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.