missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ornithine Hydrochloride, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Ignition Residue | 0.001 |
| Loss on Drying | 0.003 |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Specific Rotation | +23° to +25.0° |
| Percent Purity | 0.99 |
| Grade | Ungraded |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | L-Ornithine Hydrochloride |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.