missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride, 99%, Spectrum™ Chemical

Catalog No. 18611374
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611374 25 g Poly Bottle
18611373 100 g Poly Bottle
18611377 500 g Poly Bottle
18611372 1 kg Poly Bottle
18611376 5 kg Poly Pail
18611375 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611374 Supplier Spectrum Chemical Mfg Cor Supplier No. OR13025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +23° to +25.0°
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.