missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ornithine Dihydrochloride 98.0+%, TCI America™
$55.92 - $160.30
Chemical Identifiers
| CAS | 6211-16-1 |
|---|---|
| Molecular Formula | C5H14Cl2N2O2 |
| Molecular Weight (g/mol) | 205.08 |
| MDL Number | MFCD00066571 |
| InChI Key | HGBAVEGDXFHRQP-FHNDMYTFSA-N |
| Synonym | H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic Acid Dihydrochloride, (S)-2,5-Diaminovaleric Acid Dihydrochloride |
| PubChem CID | 80341 |
| IUPAC Name | dihydrogen (2S)-2,5-diaminopentanoic acid dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].NCCC[C@H](N)C(O)=O |
Chemical Identifiers
| 6211-16-1 | |
| 205.08 | |
| HGBAVEGDXFHRQP-FHNDMYTFSA-N | |
| 80341 | |
| [H+].[H+].[Cl-].[Cl-].NCCC[C@H](N)C(O)=O |
| C5H14Cl2N2O2 | |
| MFCD00066571 | |
| H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic Acid Dihydrochloride, (S)-2,5-Diaminovaleric Acid Dihydrochloride | |
| dihydrogen (2S)-2,5-diaminopentanoic acid dichloride |
Specifications
| 6211-16-1 | |
| C5H14Cl2N2O2 | |
| 1 g | |
| HGBAVEGDXFHRQP-FHNDMYTFSA-N | |
| dihydrogen (2S)-2,5-diaminopentanoic acid dichloride | |
| 80341 | |
| ≥98.0% (N) | |
| L-Ornithine Dihydrochloride |
| White | |
| MFCD00066571 | |
| H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic Acid Dihydrochloride, (S)-2,5-Diaminovaleric Acid Dihydrochloride | |
| [H+].[H+].[Cl-].[Cl-].NCCC[C@H](N)C(O)=O | |
| 205.08 | |
| 205.08 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1561
TSCA : Yes