missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Dihydrochloride 98.0+%, TCI America™

Catalog No. O00895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00895G 5 g
O00891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00895G Supplier TCI America Supplier No. O00895G
Only null left

Chemical Identifiers

CAS 6211-16-1
Molecular Formula C5H14Cl2N2O2
Molecular Weight (g/mol) 205.08
MDL Number MFCD00066571
InChI Key HGBAVEGDXFHRQP-FHNDMYTFSA-N
Synonym H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic Acid Dihydrochloride, (S)-2,5-Diaminovaleric Acid Dihydrochloride
PubChem CID 80341
IUPAC Name dihydrogen (2S)-2,5-diaminopentanoic acid dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 205.08
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Ornithine Dihydrochloride
EINECSNumber (9)-1561
TSCA Yes