missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Norvaline Ethyl Ester Hydrochloride 98.0+%, TCI America™
Supplier: TCI America N089825G
Specifications
| L-Norvaline Ethyl Ester Hydrochloride | |
| 109°C | |
| C7H16ClNO2 | |
| 25 g | |
| CHAJBHNQIZAJJM-UHFFFAOYNA-N | |
| ethyl 2-aminopentanoate hydrochloride | |
| 12631224 | |
| ≥98.0% (N,T) |
| 40918-51-2 | |
| White | |
| MFCD08458628 | |
| (S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl | |
| Cl.CCCC(N)C(=O)OCC | |
| 181.66 | |
| 181.66 | |
| Crystalline Powder |
Chemical Identifiers
| 40918-51-2 | |
| 181.66 | |
| CHAJBHNQIZAJJM-UHFFFAOYNA-N | |
| 12631224 | |
| Cl.CCCC(N)C(=O)OCC |
| C7H16ClNO2 | |
| MFCD08458628 | |
| (S)-2-Aminopentanoic Acid Ethyl Ester Hydrochloride, (S)-2-Aminovaleric Acid Ethyl Ester Hydrochloride, Ethyl (S)-2-Aminopentanoate Hydrochloride, Ethyl (S)-2-Aminovalerate Hydrochloride, H-Nva-OEt.HCl | |
| ethyl 2-aminopentanoate hydrochloride |
Safety and Handling
TSCA : No