missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Norleucine 99.0+%, TCI America™
$32.72 - $32.72
Chemical Identifiers
| CAS | 327-57-1 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00064423 |
| InChI Key | LRQKBLKVPFOOQJ-YFKPBYRVSA-N |
| Synonym | l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate |
| PubChem CID | 21236 |
| ChEBI | CHEBI:18347 |
| IUPAC Name | (2S)-2-aminohexanoic acid |
| SMILES | CCCC[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N0303100MG
|
TCI America
N0303100MG |
100 mg |
Each for $32.72
|
|
|||||
Chemical Identifiers
| 327-57-1 | |
| 131.18 | |
| LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
| 21236 | |
| (2S)-2-aminohexanoic acid |
| C6H13NO2 | |
| MFCD00064423 | |
| l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate | |
| CHEBI:18347 | |
| CCCC[C@H](N)C(O)=O |
Specifications
| 327-57-1 | |
| C6H13NO2 | |
| 100 mg | |
| LRQKBLKVPFOOQJ-YFKPBYRVSA-N | |
| (2S)-2-aminohexanoic acid | |
| 21236 | |
| 131.18 | |
| Crystalline Powder |
| White | |
| MFCD00064423 | |
| l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate | |
| CCCC[C@H](N)C(O)=O | |
| 131.18 | |
| CHEBI:18347 | |
| ≥99.0% (T) | |
| L-Norleucine |
Safety and Handling
RTECSNumber : RC6308000
TSCA : Yes