missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Norleucine, 99%

Catalog No. AAL0391306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0391306 5 g
AAL0391303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0391306 Supplier Thermo Scientific Chemicals Supplier No. L0391306

CAS: 327-57-1 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 327-57-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064423
InChI Key LRQKBLKVPFOOQJ-YFKPBYRVSA-N
Synonym l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate
PubChem CID 21236
ChEBI CHEBI:18347
IUPAC Name (2S)-2-aminohexanoic acid
SMILES CCCC[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721750
Merck Index 14,6706
Quantity 5 g
Formula Weight 131.18
Optical Rotation +23° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material L-(+)-Norleucine
EINECSNumber 206-321-4
RTECSNumber RC6308000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only