missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Norleucine, 99+%

Catalog No. p-3776601
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226040010 1 g Glass Bottle
AC226040050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226040010 Supplier Thermo Scientific Chemicals Supplier No. 226040010

CAS: 327-57-1 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 327-57-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064423
InChI Key LRQKBLKVPFOOQJ-YFKPBYRVSA-N
Synonym l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate
PubChem CID 21236
ChEBI CHEBI:18347
IUPAC Name (2S)-2-aminohexanoic acid
SMILES CCCC[C@H](N)C(O)=O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Loss on Drying 1% max. (105°C, 4 hrs)
Assay Percent Range 99+%
Other Amino Acids (TLC) not detectable
Packaging Glass Bottle
Specific Rotation + 22.00
Beilstein 04, 432
Merck Index 15, 6795
Sulfated Ash 0.5% max.
Solubility Information Solubility in water: 1.6 g/100mL (23 c).
Sublimation Point 0275.0°C
Specific Rotation Condition +22° (20°C c=2,6N HCl)
Quantity 1 g
Formula Weight 131.17
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Norleucine
Show More Show Less

RUO – Research Use Only