missing translation for 'onlineSavingsMsg'
Learn More

L-Noradrenaline Bitartrate Monohydrate 98.0+%, TCI America™

Catalog No. a09061g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A09061G Supplier TCI America Supplier No. A09061G
Only null left

Chemical Identifiers

CAS 108341-18-0
Molecular Formula C12H19NO10
Molecular Weight (g/mol) 337.281
MDL Number MFCD00036384
InChI Key LNBCGLZYLJMGKP-PYYOGZMQSA-N
Synonym L-Arterenol Bitartrate, L-Norepinephrine Bitartrate
PubChem CID 71311448
IUPAC Name 4-[(1S)-2-amino-1-hydroxyethyl]benzene-1,2-diol;(2R,3R)-2,3-dihydroxybutanedioic acid;hydrate
SMILES C1=CC(=C(C=C1C(CN)O)O)O.C(C(C(=O)O)O)(C(=O)O)O.O

Specifications

Melting Point 129°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 319.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Noradrenaline Bitartrate Monohydrate
RTECSNumber DN6750000
TSCA No