missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Nicotine, 99+%

Catalog No. ac181420250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181420250 25 g Glass bottle
AC181420050 5 g Glass bottle
AC181421000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC181420250 Supplier Thermo Scientific Chemicals Supplier No. 181420250
Only null left

L-Nicotine, CAS # 54-11-5, has been shown in laboratory studies to alleviate smoking habits. It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine. | CAS: 54-11-5 | C10H14N2 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
  • It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine

Applications

  • L-Nicotine has been shown in studies to alleviate smoking habits
  • It has also been shown to enhance cognitive functions such as memory
  • Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.23
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCCC1C2=CN=CC=C2

Specifications

Melting Point -80.0°C
Color Brown or Colorless to Yellow
Density 1.0150g/mL
Boiling Point 247.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5255 to 1.5275
Quantity 25 g
Beilstein 23,I,3
Merck Index 15,6609
Specific Gravity 1.015
Specific Rotation Condition −166.00 (20.00°C c=neat)
Specific Rotation − 166.00
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol,chloroform,ether,kerosene,,petrouleum ether and oils
Formula Weight 162.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material L-Nicotine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
EINECSNumber 200-193-3

RUO – Research Use Only