missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine 99.0+%, TCI America™

Catalog No. M009925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M009925G 25 g
M0099100G 100 g
M0099500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M009925G Supplier TCI America Supplier No. M009925G
Only null left

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 284°C
Color White
Quantity 25 g
Formula Weight 149.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Methionine
EINECSNumber (2)-1254
RTECSNumber PD0457000
TSCA Yes