missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoximine, 98+%

CAS: 15985-39-4 | C5H12N2O3S | 180.22 g/mol

Supplier:  Thermo Scientific Chemicals 227200050

 View more versions of this product

Catalog No. AC227200050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Methionine sulfoximine
15985-39-4
White
98+%
C5H12N2O3S
MFCD00002621
+13.50° (20°C c=1,H2O)
l-methionine sulfoximine, 2s-2-amino-4-s-methylsulfonimidoyl butanoic acid, l-methionine r,s-sulfoximine, methionine sulfoximine l, l-s-3-amino-3-carboxypropyl-s-methylsulfoximine, butanoic acid, 2-amino-4-s-methylsulfonimidoyl-, 2s, l-methionine-s,r-sulfoximine, prestwick_805, l-methionine-sulfoximine
SXTAYKAGBXMACB-DPVSGNNYSA-N
(2S)-2-amino-4-(methylsulfonimidoyl)butanoic acid
89034
180.22
Solid
98+%
210.0°C
Authentic
Glass bottle
CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
5 g
+ 13.50
Solubility in water: soluble.
CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
180.22
CHEBI:28490
98+%

Chemical Identifiers

15985-39-4
180.22
SXTAYKAGBXMACB-DPVSGNNYSA-N
89034
(2S)-2-amino-4-(methylsulfonimidoyl)butanoic acid
C5H12N2O3S
MFCD00002621
l-methionine sulfoximine, 2s-2-amino-4-s-methylsulfonimidoyl butanoic acid, l-methionine r,s-sulfoximine, methionine sulfoximine l, l-s-3-amino-3-carboxypropyl-s-methylsulfoximine, butanoic acid, 2-amino-4-s-methylsulfonimidoyl-, 2s, l-methionine-s,r-sulfoximine, prestwick_805, l-methionine-sulfoximine
CHEBI:28490
CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only