missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoximine, 98+%

Catalog No. AC227202500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227202500 250 mg Glass bottle
AC227200010 1 g Glass bottle
AC227200050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227202500 Supplier Thermo Scientific Chemicals Supplier No. 227202500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15985-39-4 | C5H12N2O3S | 180.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15985-39-4
Molecular Formula C5H12N2O3S
Molecular Weight (g/mol) 180.22
MDL Number MFCD00002621
InChI Key SXTAYKAGBXMACB-DPVSGNNYSA-N
Synonym l-methionine sulfoximine, 2s-2-amino-4-s-methylsulfonimidoyl butanoic acid, l-methionine r,s-sulfoximine, methionine sulfoximine l, l-s-3-amino-3-carboxypropyl-s-methylsulfoximine, butanoic acid, 2-amino-4-s-methylsulfonimidoyl-, 2s, l-methionine-s,r-sulfoximine, prestwick_805, l-methionine-sulfoximine
PubChem CID 89034
ChEBI CHEBI:28490
IUPAC Name (2S)-2-amino-4-(methylsulfonimidoyl)butanoic acid
SMILES CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
Quantity 250 mg
Specific Rotation Condition +13.50° (20°C c=1,H2O)
Specific Rotation + 13.50
Solubility Information Solubility in water: soluble.
Formula Weight 180.22
Percent Purity 98+%
Physical Form Solid
Chemical Name or Material L-Methionine sulfoximine, 98+%
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

RUO – Research Use Only