missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoxide

CAS: 3226-65-1 | C5H11NO3S | 165.21 g/mol

$86.86 - $1295.74

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00063093
InChI Key QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Synonym l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
PubChem CID 158980
ChEBI CHEBI:17016
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6287303
View Documents
Thermo Scientific Chemicals
J6287303
1 g
Each for $86.86
Only null left
 
AAJ6287306
View Documents
Thermo Scientific Chemicals
J6287306
5 g
Each for $326.58
Only null left
 
AAJ6287314
View Documents
Thermo Scientific Chemicals
J6287314
25 g
Each for $1,295.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3226-65-1
165.21
QEFRNWWLZKMPFJ-YGVKFDHGSA-N
158980
(2S)-2-amino-4-methylsulfinylbutanoic acid
C5H11NO3S
MFCD00063093
l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
CHEBI:17016
CS(=O)CC[C@H](N)C(O)=O

Specifications

3226-65-1
C5H11NO3S
4129939
QEFRNWWLZKMPFJ-YGVKFDHGSA-N
(2S)-2-amino-4-methylsulfinylbutanoic acid
165.21
CHEBI:17016
Powder
White
MFCD00063093
l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
CS(=O)CC[C@H](N)C(O)=O
1 g
158980
165.21
L-Methionine sulfoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 221-758-0

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only