missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ L-Methionine, Calbiochem™,

Supplier:  MilliporeSigma™ 4500100GM

Catalog No. 4500100GM


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

63-68-3
C5H11NO2S
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
149.21
CHEBI:16643
Crystalline Solid
Colorless to White
Glass Bottle
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
CSCC[C@H](N)C(O)=O
100 g
6137
≥98.5% (Assay)

Chemical Identifiers

63-68-3
149.21
FFEARJCKVFRZRR-BYPYZUCNSA-N
6137
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
C5H11NO2S
MFCD00063097 MFCD00801344
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
CHEBI:16643
CSCC[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions