missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Catalog No. p-7080685
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031822 100 g
AAA1031836 500 g
AAA103180E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1031822 Supplier Thermo Scientific Chemicals Supplier No. A1031822
Only null left

L-Methionine (CAS # 63-68-3) is an essential amino acid that contains sulphur. L-Methionine is involved in cellular detoxification in several species. | CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

L-Methionine iserves as a chelating agent for heavy metals, as a flavoring agent and nutritional supplement for foods. It acts as a feed additive, vegetable oil enrichment and as a single cell protein. In addition to this, it is used as a hepatoprotectant and also acts as a lipotropic agent and prevents excess fat buildup in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Methionine is an essential amino acid required for tissue repair and maintenance

• L-Methionine affects absorption of selenium and zinc

• L-Methionine can assist cellular detoxification by chelating lead and mercury

• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver

Application

• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications

• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water and hydrochloride. Slightly soluble in acetic acid. Insoluble in ethanol,benzene,acetone and ether.
Quantity 100 g
Formula Weight 149.21
Optical Rotation +23° (c=2 in 2N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Methionine
Hazard Category H500
EINECSNumber 200-562-9
RTECSNumber PD0457000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only