missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Catalog No. AC166165000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166165000 500 g Plastic Bottle
AC166160250 25 g Plastic Bottle
AC166161000 100 g Plastic Bottle
AC166160025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC166165000 Supplier Thermo Scientific Chemicals Supplier No. 166165000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 276.0°C to 279.0°C
Ammonium 300ppm max.
pH 5.5 to 6.5 (aq. soln. 1 in 100)
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 15ppm max.
Individual Impurity 0.5% max. of any individual impurity
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Loss on Drying 0.5% max. (1 g, 105°C)
Assay Percent Range 98+%
Packaging Plastic Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Specific Rotation + 23.25
Beilstein 04, II, 938
Sulfate 300ppm max.
Merck Index 15, 6047
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble in water: 53.7g/L (20°C). Other solubilities: insoluble in abs.alcohol, ether, petr.ether,, benzene,acetone, ethanol, diethylether, soluble in mineral acids, soluble in warm, dilute alcohol
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Specific Rotation Condition +23.25° (20°C c=2,6N HCl)
Quantity 500 g
Formula Weight 149.21
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Methionine, 98+%
Show More Show Less
Hazard Category AIR SENSITIVE
EINECSNumber 200-562-9
RTECSNumber PD0457000
TSCA TSCA

RUO – Research Use Only