Learn More
L-Menthone, 97%
CAS: 14073-97-3 | C10H18O | 154.25 g/mol
Supplier: Thermo Scientific Chemicals A1367922
| Quantity | 100 g |
|---|
Description
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
Solubility
Soluble in ether, alcohol, benzene, acetone.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 14073-97-3 | |
| 154.25 | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| 26447 | |
| (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one |
| C10H18O | |
| MFCD00001634,MFCD00136033 | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| CHEBI:15410 | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O |
Specifications
| L-Menthone | |
| 0.893 | |
| 72°C (162°F) | |
| C10H18O | |
| 100 g | |
| 3648743 | |
| --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one | |
| NFLGAXVYCFJBMK-BDAKNGLRSA-N | |
| −25° (Neat) | |
| 154.25 | |
| CHEBI:15410 | |
| 97% |
| 14073-97-3 | |
| 208°C to 210°C | |
| 97% | |
| 1.45 | |
| MFCD00001634,MFCD00136033 | |
| 14,5838 | |
| Soluble in ether,alcohol,benzene,acetone. | |
| CC(C)[C@@H]1CC[C@@H](C)CC1=O | |
| (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one | |
| 26447 | |
| 154.25 | |
| Liquid |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 237-926-1
RTECSNumber : OS9542200
TSCA : Yes
Recommended Storage : Ambient temperatures