missing translation for 'onlineSavingsMsg'
Learn More

L-Menthone, 97%

Catalog No. AAA1367909
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1367909 10 g
AAA1367918 50 g
AAA1367922 100 g
AAA1367930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1367909 Supplier Thermo Scientific Chemicals Supplier No. A1367909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14073-97-3 | C10H18O | 154.25 g/mol

L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

Solubility
Soluble in ether, alcohol, benzene, acetone.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 14073-97-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001634,MFCD00136033
InChI Key NFLGAXVYCFJBMK-BDAKNGLRSA-N
Synonym --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
PubChem CID 26447
ChEBI CHEBI:15410
IUPAC Name (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

Density 0.893
Boiling Point 208°C to 210°C
Flash Point 72°C (162°F)
Assay Percent Range 97%
Refractive Index 1.45
Quantity 10 g
Beilstein 3648743
Merck Index 14,5838
Solubility Information Soluble in ether,alcohol,benzene,acetone.
Optical Rotation −25° (Neat)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material L-Menthone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 237-926-1
RTECSNumber OS9542200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only