missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Mandelic Acid 99.0+%, TCI America™
Supplier: TCI America M0661250G
Specifications
| L-(+)-Mandelic Acid | |
| 134°C | |
| White to Yellow | |
| MFCD00004495 | |
| IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
| (2S)-2-hydroxy-2-phenylacetic acid | |
| 439616 | |
| 152.15 | |
| 250 g |
| 2.3 (10 g/L aq. soln.) | |
| 17199-29-0 | |
| C8H8O3 | |
| s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
| C1=CC=C(C=C1)C(C(=O)O)O | |
| 152.149 | |
| CHEBI:32800 | |
| ≥99.0% (GC,T) | |
| Crystalline Powder |
Chemical Identifiers
| 17199-29-0 | |
| 152.149 | |
| IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
| 439616 | |
| (2S)-2-hydroxy-2-phenylacetic acid |
| C8H8O3 | |
| MFCD00004495 | |
| s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
| CHEBI:32800 | |
| C1=CC=C(C=C1)C(C(=O)O)O |
Safety and Handling
EINECSNumber : (3)-1764
TSCA : Yes