missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Mandelic Acid 99.0+%, TCI America™

Catalog No. M066125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M066125G 25 g
M0661250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M066125G Supplier TCI America Supplier No. M066125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004495
InChI Key IWYDHOAUDWTVEP-ZETCQYMHSA-N
Synonym s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
PubChem CID 439616
ChEBI CHEBI:32800
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

pH 2.3 (10 g/L aq. soln.)
Melting Point 134°C
Color White to Yellow
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 152.15
Percent Purity ≥99.0% (GC,T)
Chemical Name or Material L-(+)-Mandelic Acid
EINECSNumber (3)-1764
TSCA Yes