missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Malic acid, 99%
Description
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.
Solubility
Soluble in water(363g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
L-(-)-Malic acid is used as a food additive, Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid receptor agonists, 1α,25-dihydroxyvitamin D3 analogue, and phoslactomycin B.
Solubility
Soluble in water(363g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 100°C to 106°C |
| Color | White |
| Density | 1.6 |
| Flash Point | 220°C (428°F) |
| Quantity | 2.5 kg |
| Beilstein | 1723541 |
| Merck Index | 14,5707 |
| Solubility Information | Soluble in water(363g/L). |
| Optical Rotation | −26° (c=5.5 in pyridine) |
| Formula Weight | 134.09 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H318-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-601-5 |
| RTECSNumber | ON7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only