Learn More
L-(-)-Malic acid, 97%
Description
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.
Solubility
Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene
Notes
Incompatible with Bases, Oxidizing agents, Reducing agents, Alkali metals .
Specifications
Specifications
| Melting Point | 100°C to 106°C |
| Density | 1.6 |
| Quantity | 100 g |
| Flash Point | 220°C (428°F) |
| Assay Percent Range | 97% |
| Beilstein | 1723541 |
| Merck Index | 14,5707 |
| Solubility Information | Soluble in acetone, dioxane, water, methanol and ethanol; Insoluble in benzene |
| Formula Weight | 134.09 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H315-H318-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-601-5 |
| RTECSNumber | ON7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only