missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine methyl ester dihydrochloride, 99%

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol

Supplier:  Thermo Scientific Chemicals A1815722

Quantity 100 g

 View more versions of this product

Catalog No. AAA1815722


Only null left

Description

Description

L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

26348-70-9
233.13
SXZCBVCQHOJXDR-ILKKLZGPSA-N
117778
[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
C7H18Cl2N2O2
MFCD00039067
l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
methyl (2S)-2,6-diaminohexanoate;dihydrochloride

Specifications

∼200°C (decomposition)
26348-70-9
MFCD00039067
l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
SXZCBVCQHOJXDR-ILKKLZGPSA-N
methyl (2S)-2,6-diaminohexanoate;dihydrochloride
233.13
233.14
99%
L-Lysine methyl ester dihydrochloride
C7H18Cl2N2O2
5302183
Soluble in water.
[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
100 g
117778
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 247-625-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions