missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine methyl ester dihydrochloride, 99%

Catalog No. AAA1815714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1815714 25 g
AAA1815722 100 g
AAA1815736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1815714 Supplier Thermo Scientific Chemicals Supplier No. A1815714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol

L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Molecular Weight (g/mol) 233.13
MDL Number MFCD00039067
InChI Key SXZCBVCQHOJXDR-ILKKLZGPSA-N
Synonym l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
PubChem CID 117778
IUPAC Name methyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 5302183
Solubility Information Soluble in water.
Quantity 25 g
Formula Weight 233.14
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material L-Lysine methyl ester dihydrochloride
EINECSNumber 247-625-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only