missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine dihydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 413361000

Catalog No. AC413361000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L-Lysine dihydrochloride
657-26-1
0.5% max. (105°C, 3 hrs)
C6H16Cl2N2O2
NH2(CH2)4CH(NH2)COOH·2HCl
+ 14.00
l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
[H+].[H+].[Cl-].[Cl-].NCCCC[C@H](N)C(O)=O
+14.00 (18.00°C c=2,water)
219.11
219.11
99%
Authentic
99%
Glass bottle
MFCD00066389
14, 5636
JBBURJFZIMRPCZ-XRIGFGBMSA-N
(2S)-2,6-diaminohexanoic acid;dihydrochloride
100g
69567
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-518-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only