missing translation for 'onlineSavingsMsg'
Learn More

L-Leucylglycine Hydrate 98.0+%, TCI America™

Catalog No. L0032100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0032100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L0032100MG Supplier TCI America Supplier No. L0032100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 686-50-0
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.227
MDL Number MFCD00065940
InChI Key LESXFEZIFXFIQR-LURJTMIESA-N
Synonym h-leu-gly-oh, s-2-2-amino-4-methylpentanamido acetic acid, l-leucyl-glycine, l-leucylglycine, leu-gly, n-l-leucylglycine, leucylglycine, l-leu-gly, chembl55711, leucyl-glycine
PubChem CID 97364
ChEBI CHEBI:74534
IUPAC Name 2-[[(2S)-2-amino-4-methylpentanoyl]amino]acetic acid
SMILES CC(C)CC(C(=O)NCC(=O)O)N

Specifications

Melting Point 236°C
Color White
Quantity 100 mg
Formula Weight 188.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucylglycine Hydrate