missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Leucinol 97.0+%, TCI America™

Catalog No. p-7131835
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01375ML 5 mL
L013725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01375ML Supplier TCI America Supplier No. L01375ML
Only null left

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 mL
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material L-(+)-Leucinol