missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Leucinol 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 210°C |
| Quantity | 5 mL |
| Formula Weight | 117.19 |
| Percent Purity | ≥97.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | L-(+)-Leucinol |
Safety and Handling
| TSCA | No |