missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Leucinol, 98%

Catalog No. p-3776664
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC179970050 Supplier Thermo Scientific Chemicals Supplier No. 179970050

CAS: 7533-40-6 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO

Specifications

Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Refractive Index 1.4490 to 1.4530 (20°C, 589nm)
Specific Rotation +3° to +5° (20°C, 589nm) (c=9, ethanol)
Density 0.91g/mL
Specific Rotation Condition +4° (20°C 589nm,c=9,ethanol)
Quantity 5 g
Physical Form Liquid
Assay Percent Range 98%
Linear Formula (CH3)2CHCH2CH(NH2)CH2OH
Beilstein 04, 298
Formula Weight 117.19
Percent Purity ≥97.5%
Chemical Name or Material L(+)-Leucinol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only