missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinol, 97%

Catalog No. AAB2374514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2374514 25 g
AAB2374506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2374514 Supplier Thermo Scientific Chemicals Supplier No. B2374514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7533-40-6 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO

Specifications

Refractive Index 1.451
Density 0.917
Boiling Point 198°C to 200°C
Quantity 25 g
Flash Point 90°C (194°F)
Optical Rotation +1.1° (Neat)
Sensitivity Air Sensitive
Beilstein 1719240
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material L-Leucinol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
EINECSNumber 231-400-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only