missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP385100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP385100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP385100 Supplier Fisher BioReagents Supplier No. FLBP385 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This amino acid is suitable for use in tissue culture systems requiring additives.

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point 286°C
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 5.5 to 6.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.2% max.
Other Amino Acids Chromatographically not detectable
Packaging Glass Bottle
Specific Rotation 14.9 to 16.1° (+ or -)
Merck Index 15, 5503
State of Solution (%T) 98% min.
Alpha Vector Leucine
Quantity 100 g
Formula Weight 131.17
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Leucine
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Moisture sensitive. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Do not ingest. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. Move to fresh air. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1
Recommended Storage RT