missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. l015525g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
L015525G 25 g
1 options

Catalog No. L015525G

Supplier: TCI America L015525G

Only null left

Chemical Identifiers

CAS 7517-19-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012494
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
PubChem CID 2723695
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 150°C
Color White
Quantity 25 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucine Methyl Ester Hydrochloride