missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Leucine methyl ester hydrochloride, 98%

Catalog No. AC125130250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125130250 Supplier Thermo Scientific Chemicals Supplier No. 125130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7517-19-3 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7517-19-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012494
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
PubChem CID 2723695
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 145.0°C to 155.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2CH3·HCl
Quantity 25 g
Beilstein 04, I, 518
Specific Rotation Condition + 20.00 (20.00°C c=4.5,MeOH)
Specific Rotation + 20.00
Solubility Information Solubility in water: soluble in water. Other solubilities: 5% soluble in methanol
Formula Weight 181.66
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Leucine methyl ester hydrochloride, 98%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 231-375-

RUO – Research Use Only