missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. p-7022225
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1489714 25 g
AAA1489722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1489714 Supplier Thermo Scientific Chemicals Supplier No. A1489714

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7517-19-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012494
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
PubChem CID 2723695
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 149°C to 151°C
Beilstein 3595133
Quantity 25 g
Formula Weight 181.66
Optical Rotation +13° (c=2 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Leucine methyl ester hydrochloride
EINECSNumber 231-375-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only