missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603937
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603937 100 g Poly Bottle
18603938 250 g Poly Bottle
18603939 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603937 Supplier Spectrum Chemical Mfg Cor Supplier No. L1203100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

L1203, 61-90-5, C6H13NO2

Spectrum™ Chemical L-Leucine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Lead (Pb) 5 mg/kg
Loss on Drying 0.2%
Packaging Poly Bottle
Quantity 100 g
Optical Rotation +14.5° to 16.5°
Percent Purity 98.5 to 101.5%
Grade FCC