missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Leucine, Cell Culture Reagent

CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

Supplier:  Thermo Scientific Chemicals J6282430

 View more versions of this product

Catalog No. AAJ6282430


Only null left

Description

Description

In cosmetics industry it is used as antistatic agent, hair conditioning and skin conditioning. L-Leucine acts as a nutrient signal to stimulate protein synthesis. It has also activates the mammalian target of rapamycin kinase that regulates cell growth. Leucine has been used as a molecular marker in the recovery of DNA from paleontological samples for PCR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In cosmetics industry it is used as antistatic agent, hair conditioning and skin conditioning. L-Leucine acts as a nutrient signal to stimulate protein synthesis. It has also activates the mammalian target of rapamycin kinase that regulates cell growth. Leucine has been used as a molecular marker in the recovery of DNA from paleontological samples for PCR analysis.

Solubility
Soluble in water. Soluble in 1M HCl (100 mg/ml), water (24.3 mg/ml), acetic acid (10.9 mg/ml), and alcohol. Insoluble in ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

>300°C
61-90-5
C6H13NO2
1721722
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
ROHFNLRQFUQHCH-MLHKIVSYNA-N
CC(C)C[C@H](N)C(O)=O
131.18
6106
131.17
Powder
L-Leucine
White
MFCD00002617
14,5451
Soluble in water. Soluble in 1M HCl (100mg/ml),water (24.3mg/ml),acetic acid (10.9mg/ml),and alcohol. Insoluble in ether.
1.293
(2S)-2-amino-4-methylpentanoic acid
250 g
CHEBI:15603
Cell Culture Reagent

Chemical Identifiers

61-90-5
131.18
ROHFNLRQFUQHCH-MLHKIVSYNA-N
6106
(2S)-2-amino-4-methylpentanoic acid
C6H13NO2
MFCD00002617
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
CHEBI:15603
CC(C)C[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-522-0

RTECSNumber : OH2850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only