missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Leucine, 99%

L-Leucine, CAS # 61-90-5, is a branched-chain aliphatic essential amino acid in humans that is important for protein synthesis and many metabolic functions.

Supplier:  Thermo Scientific Chemicals A123110E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA123110E


Only null left

Description

Description

L-Leucine plays a vital role in hemoglobin formation, protein synthesis and metabolic functions. It assists the growth and repair of muscle and bone tissue. It is used in the treatment of amyotrophic lateral sclerosis - Lou Gehrig's disease. It prevents the breakdown of muscle proteins after trauma or severe stress and may be beneficial for individuals with phenylketonuria. It is also used as a food additive and flavor enhancer. Further, it is used to preserve muscle glycogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Leucine is an essential amino acid that has an important role in the growth and repair of muscle and bone tissue
• It functions to preserve muscle glycogen and helps to prevent the breakdown of muscle proteins after trauma or severe stress
• It is also involved in hemoglobin formation and metabolic functions. This amino acid cannot be synthesized to meet the body's needs from simpler molecules, and therefore must be provided in the diet.
• L-Leucine stimulates protein synthesis by the initiation of mRNA translation

Applications

• L-Leucine is used in the biosynthesis of proteins
• In vivo studies show that L-Leucine can be used when gene defects lead to an organism's inability to metabolize the amino acid phenylalanine
• This compound also contributes to the regulation of blood-sugar levels, growth hormone production, and wound healing
• In research experiments this compound has been used as an activator of the mammalian target of rapamycin (mTOR)

Specifications

Chemical Identifiers

61-90-5
131.18
ROHFNLRQFUQHCH-MLHKIVSYNA-N
6106
(2S)-2-amino-4-methylpentanoic acid
C6H13NO2
MFCD00002617
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
CHEBI:15603
CC(C)C[C@H](N)C(O)=O

Specifications

>300°C
61-90-5
C6H13NO2
1721722
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
ROHFNLRQFUQHCH-MLHKIVSYNA-N
1.293
2500 g
6106
+15° (c=2 in 5N HCl)
99%
L-Leucine
99%
MFCD00002617
14,5451
Soluble in water,alcohol,acetic acid and hydrochloric acid. Insoluble in ether.
CC(C)C[C@H](N)C(O)=O
(2S)-2-amino-4-methylpentanoic acid
131.18
CHEBI:15603
131.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-522-0

RTECSNumber : OH2850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions