missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure

Catalog No. p-7082823
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ18280A7 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AAJ18280A7 missing translation for 'mfr' Thermo Fisher Scientific missing translation for 'supplierNo' J18280A7
Only null left

CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2 S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Packaging Plastic Bottle
Beilstein 1721722
Density 1.293
Quantity 5 kg
Formula Weight 131.17
Percent Purity 98.5 to 101.5%
Specific Gravity 1.293
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Leucine
Show More Show Less
EINECSNumber 200-522-0
RTECSNumber OH2850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only