missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucinamide Hydrochloride 98.0+%, TCI America™
$52.19 - $165.90
Chemical Identifiers
| CAS | 10466-61-2 |
|---|---|
| Molecular Formula | C6H15ClN2O |
| Molecular Weight (g/mol) | 166.65 |
| MDL Number | MFCD00013012 |
| InChI Key | VSPSRRBIXFUMOU-XZNNNFJINA-N |
| Synonym | l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl |
| PubChem CID | 2733199 |
| IUPAC Name | (2S)-2-amino-4-methylpentanamide hydrochloride |
| SMILES | Cl.CC(C)C[C@H](N)C(N)=O |
Chemical Identifiers
| 10466-61-2 | |
| 166.65 | |
| VSPSRRBIXFUMOU-XZNNNFJINA-N | |
| 2733199 | |
| Cl.CC(C)C[C@H](N)C(N)=O |
| C6H15ClN2O | |
| MFCD00013012 | |
| l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl | |
| (2S)-2-amino-4-methylpentanamide hydrochloride |
Specifications
| 10466-61-2 | |
| White | |
| MFCD00013012 | |
| l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl | |
| Cl.CC(C)C[C@H](N)C(N)=O | |
| 166.65 | |
| 166.65 | |
| Crystalline Powder |
| 256°C | |
| C6H15ClN2O | |
| 1 g | |
| VSPSRRBIXFUMOU-XZNNNFJINA-N | |
| (2S)-2-amino-4-methylpentanamide hydrochloride | |
| 2733199 | |
| ≥98.0% (T) | |
| L-Leucinamide Hydrochloride |
Safety and Handling
TSCA : No