missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinamide Hydrochloride 98.0+%, TCI America™

Catalog No. L00941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00941G 1 g
L00945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00941G Supplier TCI America Supplier No. L00941G

Chemical Identifiers

CAS 10466-61-2
Molecular Formula C6H15ClN2O
Molecular Weight (g/mol) 166.65
MDL Number MFCD00013012
InChI Key VSPSRRBIXFUMOU-XZNNNFJINA-N
Synonym l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl
PubChem CID 2733199
IUPAC Name (2S)-2-amino-4-methylpentanamide hydrochloride
SMILES Cl.CC(C)C[C@H](N)C(N)=O

Specifications

Melting Point 256°C
Color White
Quantity 1 g
Formula Weight 166.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucinamide Hydrochloride