missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucinamide hydrochloride, 99%
CAS: 10466-61-2 | C6H15ClN2O | 166.65 g/mol
$128.86 - $128.86
Chemical Identifiers
| CAS | 10466-61-2 |
|---|---|
| Molecular Formula | C6H15ClN2O |
| Molecular Weight (g/mol) | 166.65 |
| MDL Number | MFCD00013012 |
| InChI Key | VSPSRRBIXFUMOU-XZNNNFJINA-N |
| Synonym | l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl |
| PubChem CID | 2733199 |
| IUPAC Name | (2S)-2-amino-4-methylpentanamide;hydrochloride |
| SMILES | Cl.CC(C)C[C@H](N)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC302980050
|
Thermo Scientific Chemicals
302980050 |
5 g | Glass bottle |
Each for $128.86
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10466-61-2 | |
| 166.65 | |
| VSPSRRBIXFUMOU-XZNNNFJINA-N | |
| 2733199 | |
| Cl.CC(C)C[C@H](N)C(N)=O |
| C6H15ClN2O | |
| MFCD00013012 | |
| l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl | |
| (2S)-2-amino-4-methylpentanamide;hydrochloride |
Specifications
| 10466-61-2 | |
| White | |
| 98.5% min. (Argentometry) | |
| C6H15ClN2O | |
| MFCD00013012 | |
| +9° (20°C c=1,H2O) | |
| l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl | |
| Cl.CC(C)C[C@H](N)C(N)=O | |
| 166.65 | |
| 166.65 | |
| Powder |
| 244°C | |
| Authentic | |
| Glass bottle | |
| (CH3)2CHCH2CH(NH2)CONH2·HCl | |
| 5 g | |
| + 9 | |
| VSPSRRBIXFUMOU-XZNNNFJINA-N | |
| (2S)-2-amino-4-methylpentanamide;hydrochloride | |
| 2733199 | |
| 99% | |
| L-Leucinamide hydrochloride, 99% |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 233-952-2
RUO – Research Use Only