missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinamide hydrochloride, 99%

Catalog No. AC302980050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302980050 5 g Glass bottle
AC302980010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302980050 Supplier Thermo Scientific Chemicals Supplier No. 302980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10466-61-2 | C6H15ClN2O | 166.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10466-61-2
Molecular Formula C6H15ClN2O
Molecular Weight (g/mol) 166.65
MDL Number MFCD00013012
InChI Key VSPSRRBIXFUMOU-XZNNNFJINA-N
Synonym l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl
PubChem CID 2733199
IUPAC Name (2S)-2-amino-4-methylpentanamide;hydrochloride
SMILES Cl.CC(C)C[C@H](N)C(N)=O

Specifications

Melting Point 244°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH(NH2)CONH2·HCl
Quantity 5 g
Specific Rotation Condition +9° (20°C c=1,H2O)
Specific Rotation + 9
Formula Weight 166.65
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Leucinamide hydrochloride, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 233-952-2

RUO – Research Use Only